Learn More
Medchemexpress LLC Adenosine 5'-monophosphate monohydrate (Standard) | 18422-05-4 | 98.9% | C10H16N5O8P | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYA0181AR
Encompass_Preferred
Adenosine 5'-monophosphate monohydrate is an analytical standard intended for research and analytical applications. It is commonly used in qualitative, quantitative, and methodological research experiments. It is also identified as an endogenous metabolite.
  • Analytical standard for research applications.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Applied in techniques such as HPLC, GC, and MS.
  • Identified as an endogenous metabolite.
Catalog No. 50-003-58893