Learn More
Medchemexpress LLC Adenosine 5'-monophosphate-13C disodium | 4578-31-8 | 99.1% | 392.18 | 1 MG
SDP
Catalog No. 5000452772
Encompass_Preferred
Adenosine 5'-monophosphate-13C (disodium) is a 13C-labeled Adenosine 5'-monophosphate. It is an orally active purine nucleotide that participates in ATP metabolism and acts as a ligand for the adenosine 2B receptor. This compound can activate AMPK in skeletal muscle, improving insulin resistance and impaired glucose metabolism, making it suitable for diabetes research.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Participates in ATP metabolism
  • Activates AMPK in skeletal muscle
  • Improves insulin resistance and impaired glucose metabolism
  • Suitable for diabetes research
Catalog No. 50-004-52772 Supplier Medchemexpress LLC Supplier No. HYW011012S1MG
Adenosine 5'-monophosphate-13C (disodium) is a 13C-labeled Adenosine 5'-monophosphate. It is an orally active purine nucleotide that participates in ATP metabolism and acts as a ligand for the adenosine 2B receptor. This compound can activate AMPK in skeletal muscle, improving insulin resistance and impaired glucose metabolism, making it suitable for diabetes research.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Participates in ATP metabolism
  • Activates AMPK in skeletal muscle
  • Improves insulin resistance and impaired glucose metabolism
  • Suitable for diabetes research