Learn More
Selleck Chemical LLC Adenosine 5'-monophosphate monohydrate 25mg 18422-05-4 5'-Adenylic acid, 5'-AMP
SDP
Supplier:  Selleck Chemical LLC S528425MG
Encompass_Preferred
Adenosine 5'-monophosphate monohydrate (5'-Adenylic acid, 5'-AMP) is an activator of a class of protein kinases known as AMP-activated protein kinase (AMPK). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8322