Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine 5′-monophosphoramidate sodium | 102029-68-5 | 5 MG
SDP

Catalog No. 5000418793
Encompass_Preferred

Adenosine 5′-monophosphoramidate sodium is an adenosine derivative that serves as an intermediate for nucleotide synthesis. This compound significantly affects the accumulation of cyclic AMP.

  • An adenosine derivative
  • Used as an intermediate for nucleotide synthesis
  • Significantly affects the accumulation of cyclic AMP
  • Purity: 96.0%
  • Appearance: Solid
  • Color: White to off-white
  • Target: Nucleoside antimetabolite/analog
  • Pathway: Cell cycle/DNA damage

Catalog No. 50-004-18793 Supplier Medchemexpress LLC Supplier No. HYN75175MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Adenosine 5′-monophosphoramidate sodium is an adenosine derivative that serves as an intermediate for nucleotide synthesis. This compound significantly affects the accumulation of cyclic AMP.

  • An adenosine derivative
  • Used as an intermediate for nucleotide synthesis
  • Significantly affects the accumulation of cyclic AMP
  • Purity: 96.0%
  • Appearance: Solid
  • Color: White to off-white
  • Target: Nucleoside antimetabolite/analog
  • Pathway: Cell cycle/DNA damage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.