Learn More
Medchemexpress LLC Adenosine 5′-monophosphoramidate sodium | 102029-68-5 | 96.0% | 368.22 | 10 MG
SDP
Catalog No. 5000365092
Encompass_Preferred
Adenosine 5′-monophosphoramidate sodium is an adenosine derivative used as an intermediate for nucleotide synthesis. It significantly affects the accumulation of cyclic AMP.
  • An adenosine derivative
  • Can be used as an intermediate for nucleotide synthesis
  • Significantly affects the accumulation of cyclic AMP
Catalog No. 50-003-65092 Supplier Medchemexpress LLC Supplier No. 10MGHYN7517
Adenosine 5′-monophosphoramidate sodium is an adenosine derivative used as an intermediate for nucleotide synthesis. It significantly affects the accumulation of cyclic AMP.
  • An adenosine derivative
  • Can be used as an intermediate for nucleotide synthesis
  • Significantly affects the accumulation of cyclic AMP