Learn More
Medchemexpress LLC Adenosine 5'-O-(2-thiodiphosphate) trilithium salt | 73536-95-5 | MFCD00077314 | 94.9% | C10H12Li3N5O9P2S | 10MG
SDP
Catalog No. 5000262052
Encompass_Preferred
ADP-β-S trilithium is the trilithium salt of adenosine 5'-β-thiodiphosphate, used in research as a P2Y12 receptor activator. It is supplied as a white to off-white solid (CAS 73536-95-5) with listed purity ~94.9% and molecular formula C10H12Li3N5O9P2S. Store sealed and away from moisture; in solution store at -80°C for long-term stability.
  • Activator of P2Y12 receptor in cellular assays.
  • Upregulates IL-1β and IL-6 production in microglial models.
  • Promotes NF-κB phosphorylation and nuclear translocation.
  • Enhances NLRP3 inflammasome activation in relevant studies.
  • Provided as a solid suitable for preparation of assay solutions.
  • Stable under recommended sealed, low-temperature storage conditions.
Catalog No. 50-002-62052 Supplier Medchemexpress LLC Supplier No. HY134353B10MG
ADP-β-S trilithium is the trilithium salt of adenosine 5'-β-thiodiphosphate, used in research as a P2Y12 receptor activator. It is supplied as a white to off-white solid (CAS 73536-95-5) with listed purity ~94.9% and molecular formula C10H12Li3N5O9P2S. Store sealed and away from moisture; in solution store at -80°C for long-term stability.
  • Activator of P2Y12 receptor in cellular assays.
  • Upregulates IL-1β and IL-6 production in microglial models.
  • Promotes NF-κB phosphorylation and nuclear translocation.
  • Enhances NLRP3 inflammasome activation in relevant studies.
  • Provided as a solid suitable for preparation of assay solutions.
  • Stable under recommended sealed, low-temperature storage conditions.