Learn More
Medchemexpress LLC Adenosine 5'-phosphosulfate (disodium) | 102029-95-8 | 97.0% | 471.25 | 1 MG
SDP
Catalog No. 5000436445
Encompass_Preferred
Adenosine 5'-phosphosulfate disodium is a metabolism intermediate of sulfur. It can be used to study sulfotransferases and sulforeductases in various organisms.
  • For research use only.
  • Not sold to patients.
Catalog No. 50-004-36445 Supplier Medchemexpress LLC Supplier No. HY1136061MG
Adenosine 5'-phosphosulfate disodium is a metabolism intermediate of sulfur. It can be used to study sulfotransferases and sulforeductases in various organisms.
  • For research use only.
  • Not sold to patients.