Learn More
Medchemexpress LLC Adenosine 5'-β-thiodiphosphate trilithium salt | 73536-95-5 | MFCD00077314 | 94.9% | 461.07 g·mol⁻¹ | C10H12Li3N5O9P2S | 5 MG
SDP
Catalog No. 5000222026
Encompass_Preferred
ADP-β-S trilithium is the trilithium salt form of adenosine 5'-β-thiodiphosphate used as a research reagent to activate P2Y12 receptors and study purinergic signaling and inflammatory pathways. It modulates cytokine production and NF-κB/NLRP3 signaling, and is provided as a dry powder for laboratory use.
  • Activates P2Y12 receptor to modulate purinergic signaling.
  • Increases IL-1β and IL-6 production in microglial cells.
  • Promotes NF-κB phosphorylation and nuclear translocation.
  • Enhances NLRP3 inflammasome activation.
  • Supplied with stated purity and molecular data for experimental planning.
  • Suitable for biochemical and cellular assays requiring receptor activation.
Catalog No. 50-002-22026 Supplier Medchemexpress LLC Supplier No. HY134353B5MG
ADP-β-S trilithium is the trilithium salt form of adenosine 5'-β-thiodiphosphate used as a research reagent to activate P2Y12 receptors and study purinergic signaling and inflammatory pathways. It modulates cytokine production and NF-κB/NLRP3 signaling, and is provided as a dry powder for laboratory use.
  • Activates P2Y12 receptor to modulate purinergic signaling.
  • Increases IL-1β and IL-6 production in microglial cells.
  • Promotes NF-κB phosphorylation and nuclear translocation.
  • Enhances NLRP3 inflammasome activation.
  • Supplied with stated purity and molecular data for experimental planning.
  • Suitable for biochemical and cellular assays requiring receptor activation.