Learn More
Medchemexpress LLC Adenosine 5'-triphosphate-15N5 ammonium | C10H28N415N5O13P3 | 58 UG
SDP
Catalog No. 5000375725
Encompass_Preferred
Adenosine 5'-triphosphate-15N5 ammonium is the 15N labeled Adenosine 5'-triphosphate ammonium. This compound offers versatility in research applications.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-75725 Supplier Medchemexpress LLC Supplier No. HY150843S58UG
Adenosine 5'-triphosphate-15N5 ammonium is the 15N labeled Adenosine 5'-triphosphate ammonium. This compound offers versatility in research applications.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.