Learn More
Medchemexpress LLC Adenosine 5'-triphosphate-15N5,d14 dilithium | 56-65-5 | 98% | 1 MG
SDP
Catalog No. 5000410349
Encompass_Preferred
ATP-15N5,d14 (Adenosine 5'-triphosphate-15N5,d14) dilithium is a deuterium and 15N labeled version of ATP. ATP (Adenosine 5'-triphosphate) is crucial for energy storage and metabolism in living organisms. It supplies metabolic energy for driving metabolic pumps and functions as a coenzyme within cells. Additionally, ATP serves as an important endogenous signaling molecule involved in immunity and inflammation. This product is intended for research use only and is not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes (deuterium and 15N) incorporated into the molecule.
Catalog No. 50-004-10349 Supplier Medchemexpress LLC Supplier No. HYB2176S31MG
ATP-15N5,d14 (Adenosine 5'-triphosphate-15N5,d14) dilithium is a deuterium and 15N labeled version of ATP. ATP (Adenosine 5'-triphosphate) is crucial for energy storage and metabolism in living organisms. It supplies metabolic energy for driving metabolic pumps and functions as a coenzyme within cells. Additionally, ATP serves as an important endogenous signaling molecule involved in immunity and inflammation. This product is intended for research use only and is not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes (deuterium and 15N) incorporated into the molecule.