Learn More
Medchemexpress LLC Adenosine 5'-triphosphate-15N5 dilithium | 56-65-5 | 99.99% | 1 MG
SDP
Catalog No. 5000410351
Encompass_Preferred
ATP-15N5 dilithium is a 15N labeled form of Adenosine 5'-triphosphate (ATP), which is a crucial component of energy storage and metabolism in living organisms. It plays a vital role in driving metabolic pumps and functions as a coenzyme within cells. Furthermore, ATP is recognized as a significant endogenous signaling molecule involved in immunity and inflammation processes.
  • Is a 15N labeled ATP.
  • Central to energy storage and metabolism.
  • Drives metabolic pumps.
  • Functions as a coenzyme in cells.
  • Serves as an important endogenous signaling molecule in immunity and inflammation.
  • Can be used as a tracer.
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-10351 Supplier Medchemexpress LLC Supplier No. HYB2176S51MG
ATP-15N5 dilithium is a 15N labeled form of Adenosine 5'-triphosphate (ATP), which is a crucial component of energy storage and metabolism in living organisms. It plays a vital role in driving metabolic pumps and functions as a coenzyme within cells. Furthermore, ATP is recognized as a significant endogenous signaling molecule involved in immunity and inflammation processes.
  • Is a 15N labeled ATP.
  • Central to energy storage and metabolism.
  • Drives metabolic pumps.
  • Functions as a coenzyme in cells.
  • Serves as an important endogenous signaling molecule in immunity and inflammation.
  • Can be used as a tracer.
  • Suitable as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.