Learn More
Sigma Aldrich Fine Chemicals Biosciences Adenosine 5′-triphosphate disodium salt hydrate, 34369-07-8, MFCD00150755, 10 g
Catalog No. NC0630126
Encompass
Empirical Formula (Hill Notation): C10H14N5Na2O13P3 · xH2O, Molecular Weight: 551.14 (anhydrous basis), Grade I, ≥99%, from microbial, Synonym: ATP disodium salt. Adenosine 5′-triphosphate (ATP) is a central component of energy storage and metabolism in vivo. ATP is use in many cellular processes, respiration, biosynthetic reactions, motility, and cell division.
Catalog No. NC0630126 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A238310G
Empirical Formula (Hill Notation): C10H14N5Na2O13P3 · xH2O, Molecular Weight: 551.14 (anhydrous basis), Grade I, ≥99%, from microbial, Synonym: ATP disodium salt. Adenosine 5′-triphosphate (ATP) is a central component of energy storage and metabolism in vivo. ATP is use in many cellular processes, respiration, biosynthetic reactions, motility, and cell division.