Learn More
Apexbio Technology LLC Adenosine 5-triphosphate 34369-07-8 500mg
SDP
Catalog No. 5000090049
Encompass_Preferred
Adenosine 5-triphosphate (ATP CAS 34369-07-8) also known as ATP disodium salt hydrate is a nucleotide central to cellular energy transfer It functions as the primary intracellular energy carrier undergoing hydrolysis to release energy required for numerous biochemical processes including muscle contraction signal transduction and biosynthetic reactions ATP also serves as a substrate for kinases in phosphorylation events and plays a regulatory role in purinergic signaling pathways In biomedical research ATP is widely used to study cellular metabolism enzyme activities and receptor-mediated responses
Catalog No. 50-000-90049 Supplier Apexbio Technology LLC Supplier No. B7876500
Adenosine 5-triphosphate (ATP CAS 34369-07-8) also known as ATP disodium salt hydrate is a nucleotide central to cellular energy transfer It functions as the primary intracellular energy carrier undergoing hydrolysis to release energy required for numerous biochemical processes including muscle contraction signal transduction and biosynthetic reactions ATP also serves as a substrate for kinases in phosphorylation events and plays a regulatory role in purinergic signaling pathways In biomedical research ATP is widely used to study cellular metabolism enzyme activities and receptor-mediated responses