Learn More
Medchemexpress LLC Adenosine 5'-triphosphate | 56-65-5 | 95.7% | 25 MG
SDP
Catalog No. 5000412640
Encompass_Preferred
ATP (Standard) (Adenosine 5'-triphosphate (Standard)) is the analytical standard of ATP, intended for research and analytical applications. It is a central component of energy storage and metabolism in vivo, provides metabolic energy to drive metabolic pumps, and serves as a coenzyme in cells. It is also an important endogenous signaling molecule in immunity and inflammation. It is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used as an analytical standard for research and analytical applications, including HPLC, GC, and MS.
  • Synergistically activates the NLRP3 inflammasome in HGFs.
  • Induces secretion of IL-1β, KC, and MIP-2 from BMDMs in a caspase-1 activation-dependent manner.
  • Promotes neutrophil chemotaxis in vitro.
  • Protects mice against bacterial infection.
  • Induces the secretion of IL-1β, KC, and MIP-2 and neutrophils recruitment in vivo.
Catalog No. 50-004-12640 Supplier Medchemexpress LLC Supplier No. 25MGHYB2176R
ATP (Standard) (Adenosine 5'-triphosphate (Standard)) is the analytical standard of ATP, intended for research and analytical applications. It is a central component of energy storage and metabolism in vivo, provides metabolic energy to drive metabolic pumps, and serves as a coenzyme in cells. It is also an important endogenous signaling molecule in immunity and inflammation. It is commonly used in qualitative, quantitative, and methodological research experiments.
  • Used as an analytical standard for research and analytical applications, including HPLC, GC, and MS.
  • Synergistically activates the NLRP3 inflammasome in HGFs.
  • Induces secretion of IL-1β, KC, and MIP-2 from BMDMs in a caspase-1 activation-dependent manner.
  • Promotes neutrophil chemotaxis in vitro.
  • Protects mice against bacterial infection.
  • Induces the secretion of IL-1β, KC, and MIP-2 and neutrophils recruitment in vivo.