Learn More
Medchemexpress LLC Adenosine 5'-triphosphate | 56-65-5 | 95.7% | 507.18 g/mol | C10H16N5O13P3 | 10 MG
SDP
Catalog No. 5000297025
Encompass_Preferred
Adenosine 5'-triphosphate (ATP) standard supplied as a 10 mg solid for analytical and research use. It is intended as a calibration and reference material for quantitation of ATP in biochemical assays, metabolic studies, and signaling research.
  • Analytical reference standard for ATP quantitation.
  • High purity: 95.7%.
  • White to off-white solid suitable for dissolution in aqueous buffers.
  • Useful for assay calibration, method development, and quality control.
  • Includes CAS number 56-65-5 for clear substance identification.
Catalog No. 50-002-97025 Supplier Medchemexpress LLC Supplier No. HYB2176R10MG
Adenosine 5'-triphosphate (ATP) standard supplied as a 10 mg solid for analytical and research use. It is intended as a calibration and reference material for quantitation of ATP in biochemical assays, metabolic studies, and signaling research.
  • Analytical reference standard for ATP quantitation.
  • High purity: 95.7%.
  • White to off-white solid suitable for dissolution in aqueous buffers.
  • Useful for assay calibration, method development, and quality control.
  • Includes CAS number 56-65-5 for clear substance identification.