Learn More
Medchemexpress LLC Adenosine 5'-triphosphate | 56-65-5 | 99.9% | 5 G
SDP
Catalog No. 5000412317
Encompass_Preferred
ATP (Adenosine 5'-triphosphate) is a central component of energy storage and metabolism in vivo. It provides metabolic energy for pumps and acts as a coenzyme in cells. ATP is also an important endogenous signaling molecule in immunity and inflammation, capable of activating the NLRP3 inflammasome and inducing IL-1β and chemokine secretion. It exhibits anti-bacterial infection effects and can protect mice against bacterial infection.
  • Provides metabolic energy and acts as a coenzyme
  • Functions as an important endogenous signaling molecule in immunity and inflammation
  • Activates the NLRP3 inflammasome
  • Induces IL-1β and chemokine secretion
  • Exhibits anti-bacterial infection effects
  • Protects against bacterial infection in mice
  • Synergistically activates the NLRP3 inflammasome with LPS in HGFs
  • Induces secretion of IL-1β, KC, and MIP-2 from BMDMs
  • Promotes neutrophil chemotaxis
Catalog No. 50-004-12317 Supplier Medchemexpress LLC Supplier No. HYB21765G
ATP (Adenosine 5'-triphosphate) is a central component of energy storage and metabolism in vivo. It provides metabolic energy for pumps and acts as a coenzyme in cells. ATP is also an important endogenous signaling molecule in immunity and inflammation, capable of activating the NLRP3 inflammasome and inducing IL-1β and chemokine secretion. It exhibits anti-bacterial infection effects and can protect mice against bacterial infection.
  • Provides metabolic energy and acts as a coenzyme
  • Functions as an important endogenous signaling molecule in immunity and inflammation
  • Activates the NLRP3 inflammasome
  • Induces IL-1β and chemokine secretion
  • Exhibits anti-bacterial infection effects
  • Protects against bacterial infection in mice
  • Synergistically activates the NLRP3 inflammasome with LPS in HGFs
  • Induces secretion of IL-1β, KC, and MIP-2 from BMDMs
  • Promotes neutrophil chemotaxis