Learn More
Enzo Life Sciences Adenosine 5'-triphosphate . disodium salt, (5g), CAS Number: 987-65-5
SDP
Catalog No. NC1215754
Encompass_Preferred
Alternative Name: ATP . 2Na. Formula: C10H14N5O13P3 . 2Na. MW: 551.1. CAS Number: 987-65-5. MI: 14: 155. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water. Long Term Storage: -20°C
Catalog No. NC1215754 Supplier Enzo Life Sciences Supplier No. ALX480021G005
Alternative Name: ATP . 2Na. Formula: C10H14N5O13P3 . 2Na. MW: 551.1. CAS Number: 987-65-5. MI: 14: 155. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water. Long Term Storage: -20°C