Learn More
New England Biolabs, Inc. Adenosine-5 Triphosphate (ATP) – 5 ml
SDP
Supplier:  New England Biolabs, Inc. P0756L
Encompass
Adenosine 5-Triphosphate (ATP) is nucleotide used as a substrate for many ATP-dependent enzyme systems, including protein kinases, protein phosphatases and DNA ligation.
Catalog No. 50-994-923