Learn More
Abcam Adenosine 5'-triphosphate (ATP) disodium salt trihydrate, nucleotide coenzyme, 5G
Catalog No. 502704595
Encompass_Preferred
MW 605.24 g/mol, Purity >99%. Central component of energy storage and metabolism in vivo. ATP is used in many cellular processes, respiration, biosynthetic reactions, motility, and cell division. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase(s) that produce the second messenger cAMP. ATP provides the metabolic energy to drive metabolic pumps, and serves as a coenzyme in a wide array of enzymatic reactions. P₂ purinergic agonist that increases Ca²+-activated K+ channel activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4595 Supplier Abcam Supplier No. AB1465255G
MW 605.24 g/mol, Purity >99%. Central component of energy storage and metabolism in vivo. ATP is used in many cellular processes, respiration, biosynthetic reactions, motility, and cell division. ATP is a substrate of many kinases involved in cell signaling and of adenylate cyclase(s) that produce the second messenger cAMP. ATP provides the metabolic energy to drive metabolic pumps, and serves as a coenzyme in a wide array of enzymatic reactions. P₂ purinergic agonist that increases Ca²+-activated K+ channel activity.
The product is subject to the following: Abcam Restricted Use Statement