Learn More
Medchemexpress LLC Adenosine 5'-triphosphate(ATP)-d4 (ammonium salt) | 2483831-82-7 | C10H24D4N9O13P3 | 5.79 MG
SDP
Catalog No. 5000375851
Encompass_Preferred
Adenosine 5'-triphosphate(ATP)-d4 (ammonium salt) is a deuterium labeled ATP ammonium salt. It serves as a tracer and an internal standard for quantitative analysis, leveraging deuterium substitution's potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration affects pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-75851 Supplier Medchemexpress LLC Supplier No. HY150770S5.79MG
Adenosine 5'-triphosphate(ATP)-d4 (ammonium salt) is a deuterium labeled ATP ammonium salt. It serves as a tracer and an internal standard for quantitative analysis, leveraging deuterium substitution's potential to influence pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration affects pharmacokinetic and metabolic profiles of drugs.