Learn More
Medchemexpress LLC Adenosine 5'-triphosphate(ATP)-d4 ammonium solution | 2483831-82-7 | C10H24D4N9O13P3 | 11.59MG
SDP
Catalog No. 5000374816
Encompass_Preferred
Adenosine 5'-triphosphate(ATP)-d4 (ammonium salt) is the deuterium labeled Adenosine 5'-triphosphate(ATP) ammonium salt. It is intended for research use only. This compound can be utilized as a tracer and as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS. Deuterium substitution, involving stable heavy isotopes, is often used in drug molecules as tracers for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Deuterium labeled compound
  • For research use only
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuterium substitution may impact pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-74816 Supplier Medchemexpress LLC Supplier No. HY150770S11.59MG
Adenosine 5'-triphosphate(ATP)-d4 (ammonium salt) is the deuterium labeled Adenosine 5'-triphosphate(ATP) ammonium salt. It is intended for research use only. This compound can be utilized as a tracer and as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS. Deuterium substitution, involving stable heavy isotopes, is often used in drug molecules as tracers for quantitation during drug development and can influence pharmacokinetic and metabolic profiles.
  • Deuterium labeled compound
  • For research use only
  • Can be used as a tracer
  • Suitable as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
  • Deuterium substitution may impact pharmacokinetic and metabolic profiles of drugs