Learn More
Medchemexpress LLC Adenosine 5'-triphosphate-d14 dilithium | 56-65-5 | 99.3% | 1 MG
SDP
Catalog No. 5000410350
Encompass_Preferred
ATP-d14 dilithium (Adenosine 5'-triphosphate-d14 dilithium) is a deuterium-labeled form of ATP, a central component in energy storage and metabolism. It functions as a coenzyme in cells and plays a role as an endogenous signaling molecule in immunity and inflammation. It is suitable for use as a tracer or as an internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-10350 Supplier Medchemexpress LLC Supplier No. HYB2176S41MG
ATP-d14 dilithium (Adenosine 5'-triphosphate-d14 dilithium) is a deuterium-labeled form of ATP, a central component in energy storage and metabolism. It functions as a coenzyme in cells and plays a role as an endogenous signaling molecule in immunity and inflammation. It is suitable for use as a tracer or as an internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.