Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Adenosine 5'-Triphosphate Disodium Salt Hydrate

Catalog No. A2620950G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
50 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2620950G 50 g
A262091G 1 g
A262095G 5 g
A2620910G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A2620950G Supplier Sigma Aldrich Supplier No. A2620950G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Adenosine 5'-Triphosphate Disodium Salt Hydrate

Specifications

CAS 106-95-6
Melting Point -119°C
Density 1.398 g/mL at 25°C
Boiling Point 70°C to 71°C
Molecular Formula C3H5Br
Linear Formula CH2=CHCH2Br
Quantity 50 g
MDL Number MFCD00000244
Synonym 5'-ATP-Na2; ATP disodium salt hydrate
Molecular Weight (g/mol) 120.98 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.