Learn More
Medchemexpress LLC Adenosine-5'-triphosphate disodium trihydrate | 51963-61-2 | 99.9% | 500 MG
SDP
Catalog No. 5000409958
Encompass_Preferred
ATP disodium trihydrate is a central component of energy storage and metabolism in vivo. It provides metabolic energy for metabolic pumps and acts as a coenzyme in cells. It is also an important endogenous signaling molecule in immunity and inflammation. It protects mice against bacterial infection in vivo and promotes neutrophil chemotaxis in vitro.
  • Provides metabolic energy for metabolic pumps.
  • Acts as a coenzyme in cells.
  • Important endogenous signaling molecule in immunity and inflammation.
  • Protects against bacterial infection in vivo.
  • Promotes neutrophil chemotaxis in vitro.
Catalog No. 50-004-09958 Supplier Medchemexpress LLC Supplier No. HYB2176A500MG
ATP disodium trihydrate is a central component of energy storage and metabolism in vivo. It provides metabolic energy for metabolic pumps and acts as a coenzyme in cells. It is also an important endogenous signaling molecule in immunity and inflammation. It protects mice against bacterial infection in vivo and promotes neutrophil chemotaxis in vitro.
  • Provides metabolic energy for metabolic pumps.
  • Acts as a coenzyme in cells.
  • Important endogenous signaling molecule in immunity and inflammation.
  • Protects against bacterial infection in vivo.
  • Promotes neutrophil chemotaxis in vitro.