Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine | 58-61-7 | 100.0% | 25 G
SDP

Catalog No. 5000407404
Encompass_Preferred

Adenosine is a ubiquitous and BBB-permeable endogenous autacoid. It acts through four G protein-coupled receptors: A1, A2A, A2B, and A3. This compound impacts nearly all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation. It is for research use only and not sold to patients.

  • Acts through four G protein-coupled receptors: A1, A2A, A2B, and A3
  • Impacts neuronal activity, vascular function, platelet aggregation, and blood cell regulation
  • Ubiquitous and BBB-permeable endogenous autacoid
  • Displays cytoprotective function
  • Purity: 99.96%
  • Molecular weight: 267.25
  • Appearance: Solid, white to off-white

Catalog No. 50-004-07404 Supplier Medchemexpress LLC Supplier No. 25GHYB0228
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine is a ubiquitous and BBB-permeable endogenous autacoid. It acts through four G protein-coupled receptors: A1, A2A, A2B, and A3. This compound impacts nearly all aspects of cellular physiology, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation. It is for research use only and not sold to patients.

  • Acts through four G protein-coupled receptors: A1, A2A, A2B, and A3
  • Impacts neuronal activity, vascular function, platelet aggregation, and blood cell regulation
  • Ubiquitous and BBB-permeable endogenous autacoid
  • Displays cytoprotective function
  • Purity: 99.96%
  • Molecular weight: 267.25
  • Appearance: Solid, white to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.