Learn More
Apexbio Technology LLC Adenosine 58-61-7 5g
SDP
Catalog No. 5000090308
Encompass_Preferred
Adenosine (CAS 58-61-7) is a purine nucleoside consisting of adenine covalently linked to a ribose sugar via a -N9-glycosidic bond Endogenously adenosine modulates cellular signaling through activation of specific G protein-coupled adenosine receptors (A1 A2A A2B and A3) influencing diverse physiological processes such as neurotransmission vasodilation and immune responses In biomedical research adenosine is widely utilized to investigate purinergic signaling pathways cardiac function and the regulation of inflammatory and neuroprotective mechanisms as well as serving as a pharmacological tool in receptor binding and functional assays
Catalog No. 50-000-90308 Supplier Apexbio Technology LLC Supplier No. B18775000
Adenosine (CAS 58-61-7) is a purine nucleoside consisting of adenine covalently linked to a ribose sugar via a -N9-glycosidic bond Endogenously adenosine modulates cellular signaling through activation of specific G protein-coupled adenosine receptors (A1 A2A A2B and A3) influencing diverse physiological processes such as neurotransmission vasodilation and immune responses In biomedical research adenosine is widely utilized to investigate purinergic signaling pathways cardiac function and the regulation of inflammatory and neuroprotective mechanisms as well as serving as a pharmacological tool in receptor binding and functional assays