Learn More
Medchemexpress LLC Adenosine | 58-61-7 | 99.5% | 10 G
SDP
Catalog No. 5000405742
Encompass_Preferred
Adenosine (Adenine riboside) is a ubiquitous and BBB-permeable endogenous autacoid. It acts through four G protein-coupled receptors: A1, A2A, A2B, and A3. Adenosine impacts various cellular physiological processes, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.
  • Ubiquitous endogenous autacoid.
  • BBB-permeable.
  • Acts via A1, A2A, A2B, and A3 G protein-coupled receptors.
  • Affects neuronal activity.
  • Influences vascular function.
  • Impacts platelet aggregation.
  • Regulates blood cell function.
  • Relevant to apoptosis research.
  • Relevant to autophagy research.
  • Considered an endogenous metabolite.
  • Used in metabolic disease research.
  • Used in cancer research.
  • Classified as a nucleoside antimetabolite/analog.
Catalog No. 50-004-05742 Supplier Medchemexpress LLC Supplier No. HYB022810G
Adenosine (Adenine riboside) is a ubiquitous and BBB-permeable endogenous autacoid. It acts through four G protein-coupled receptors: A1, A2A, A2B, and A3. Adenosine impacts various cellular physiological processes, including neuronal activity, vascular function, platelet aggregation, and blood cell regulation.
  • Ubiquitous endogenous autacoid.
  • BBB-permeable.
  • Acts via A1, A2A, A2B, and A3 G protein-coupled receptors.
  • Affects neuronal activity.
  • Influences vascular function.
  • Impacts platelet aggregation.
  • Regulates blood cell function.
  • Relevant to apoptosis research.
  • Relevant to autophagy research.
  • Considered an endogenous metabolite.
  • Used in metabolic disease research.
  • Used in cancer research.
  • Classified as a nucleoside antimetabolite/analog.