Learn More
Medchemexpress LLC Adenosine, 8-[(4-chlorophenyl)thio]-, cyclic 3',5'-(hydrogen phosphate), sodium salt | 93882-12-3 | 99.9% | 493.79 | 1 ML
SDP
Catalog No. 5000438753
Encompass_Preferred
8-CPT-Cyclic AMP (8-CPT-cAMP) sodium is a selective activator of cyclic AMP-dependent protein kinase (PKA). It is also a potent inhibitor of the cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM, and inhibits PDE III and PDE IV with IC50Epac. 8-CPT-Cyclic AMP sodium is also a potent Epac activator. It has been shown to delay TNF-α/CHX-induced and α-Fas-induced apoptosis in neutrophils at a concentration of 0.7 mM, being more efficient against apoptosis induced by TNF-α/CHX.
  • Selective activator of cyclic AMP-dependent protein kinase (PKA).
  • Potent inhibitor of cyclic GMP-specific phosphodiesterase (PDE VA).
  • Inhibits PDE III and PDE IV.
  • Potent Epac activator.
  • Can delay TNF-α/CHX-induced and α-Fas-induced apoptosis in neutrophils.
Catalog No. 50-004-38753 Supplier Medchemexpress LLC Supplier No. HY11167310MM/1ML
8-CPT-Cyclic AMP (8-CPT-cAMP) sodium is a selective activator of cyclic AMP-dependent protein kinase (PKA). It is also a potent inhibitor of the cyclic GMP-specific phosphodiesterase (PDE VA) with an IC50 of 0.9 μM, and inhibits PDE III and PDE IV with IC50Epac. 8-CPT-Cyclic AMP sodium is also a potent Epac activator. It has been shown to delay TNF-α/CHX-induced and α-Fas-induced apoptosis in neutrophils at a concentration of 0.7 mM, being more efficient against apoptosis induced by TNF-α/CHX.
  • Selective activator of cyclic AMP-dependent protein kinase (PKA).
  • Potent inhibitor of cyclic GMP-specific phosphodiesterase (PDE VA).
  • Inhibits PDE III and PDE IV.
  • Potent Epac activator.
  • Can delay TNF-α/CHX-induced and α-Fas-induced apoptosis in neutrophils.