Learn More
Medchemexpress LLC Adenosine, 8-bromo-, cyclic 3',5'-[hydrogen P(S)]-phosphorothioate], sodium salt (1:1) | 1573115-90-8 | 99.9% | 446.15 | 500 UG
SDP
Catalog No. 5000351045
Encompass_Preferred
Sp-8-Br-cAMPS sodium is a cAMP analog that activates protein kinase A (PKA). It is known to inhibit T-cell proliferation and the haemocyte non-self response in Lepidoptera larvae. This compound also inhibits phagocytosis of certain bacteria and modulates cytokine expression.
  • Activates protein kinase A (PKA) with an EC50 of 360 nM
  • Inhibits T-cell proliferation
  • Inhibits haemocyte non-self response in Lepidoptera larvae
  • Inhibits phagocytosis of *X. nematophila* and *B. subtilis*
  • Modulates cytokine expression (IFN-γ, TNF-α, IL-2, IL-4)
Catalog No. 50-003-51045 Supplier Medchemexpress LLC Supplier No. HY137640A500UG
Sp-8-Br-cAMPS sodium is a cAMP analog that activates protein kinase A (PKA). It is known to inhibit T-cell proliferation and the haemocyte non-self response in Lepidoptera larvae. This compound also inhibits phagocytosis of certain bacteria and modulates cytokine expression.
  • Activates protein kinase A (PKA) with an EC50 of 360 nM
  • Inhibits T-cell proliferation
  • Inhibits haemocyte non-self response in Lepidoptera larvae
  • Inhibits phagocytosis of *X. nematophila* and *B. subtilis*
  • Modulates cytokine expression (IFN-γ, TNF-α, IL-2, IL-4)