Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine, 8-bromo-, cyclic 3',5'-(hydrogen phosphate), sodium salt (1:1) | 76939-46-3 | MFCD00149373 | ≥98.0% | 430.08 | C10H10BrN5NaO6P | 10MM 1ML
SDP

Catalog No. 5000235533
Encompass_Preferred

8-Bromo-cAMP sodium salt is a cell-permeable cyclic AMP analog used to activate cyclic AMP-dependent protein kinase (PKA) in cellular and biochemical research. It is supplied as a solid and as a 10 mM solution in dimethyl sulfoxide (DMSO) (1 mL). The material is provided with high purity (≥98% by HPLC) and is characterized by molecular weight 430.08 and formula C10H10BrN5NaO6P. For research use only.

  • Cell-permeable cyclic AMP analog for intracellular studies.
  • Activates cyclic AMP-dependent protein kinase (PKA).
  • Available as solid or 10 mM solution in DMSO (1 mL).
  • High purity: ≥98% (HPLC).
  • Molecular weight 430.08 and formula C10H10BrN5NaO6P.
  • Suitable for studies of cAMP/PKA signaling, apoptosis, and proliferation.

Catalog No. 50-002-35533 Supplier Medchemexpress LLC Supplier No. HY1230610MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

8-Bromo-cAMP sodium salt is a cell-permeable cyclic AMP analog used to activate cyclic AMP-dependent protein kinase (PKA) in cellular and biochemical research. It is supplied as a solid and as a 10 mM solution in dimethyl sulfoxide (DMSO) (1 mL). The material is provided with high purity (≥98% by HPLC) and is characterized by molecular weight 430.08 and formula C10H10BrN5NaO6P. For research use only.

  • Cell-permeable cyclic AMP analog for intracellular studies.
  • Activates cyclic AMP-dependent protein kinase (PKA).
  • Available as solid or 10 mM solution in DMSO (1 mL).
  • High purity: ≥98% (HPLC).
  • Molecular weight 430.08 and formula C10H10BrN5NaO6P.
  • Suitable for studies of cAMP/PKA signaling, apoptosis, and proliferation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.