Learn More
Medchemexpress LLC Adenosine, 8-bromo-, cyclic 3',5'-(hydrogen phosphate), sodium salt | 76939-46-3 | MFCD00005844 | 99.9% | 430.08 g·mol⁻¹ | C10H10BrN5NaO6P | 10MG
SDP
Catalog No. 5000237579
Encompass_Preferred
8-Bromo-cAMP sodium salt is a cell-permeable cyclic adenosine monophosphate (cAMP) analog used to activate cAMP-dependent protein kinase (PKA) in biochemical and cell-based studies. Supplied as a white to off-white solid with high reported purity, it is suitable for probing PKA signaling and for experiments requiring a stable cAMP mimic. Store sealed, away from moisture; storage recommendations differ for solutions versus solid form.
Catalog No. 50-002-37579 Supplier Medchemexpress LLC Supplier No. HY1230610MG
8-Bromo-cAMP sodium salt is a cell-permeable cyclic adenosine monophosphate (cAMP) analog used to activate cAMP-dependent protein kinase (PKA) in biochemical and cell-based studies. Supplied as a white to off-white solid with high reported purity, it is suitable for probing PKA signaling and for experiments requiring a stable cAMP mimic. Store sealed, away from moisture; storage recommendations differ for solutions versus solid form.