Learn More
Sigma Aldrich Fine Chemicals Biosciences Adenosine 99100G
Catalog No. 502903695
Encompass
In the extracellular space ecto-5AE-nucleotidase (CD73) dephosphorylates adenosine triphosphate (ATP) to produce adenosine. Adenosine has four receptors namely A1R A2AR A2BR and A3R. Adenosine plays a key role in the osteogenic differentiation. A1R induces osteoclast differentiation and A2AR induces osteoblast differentiation.
Catalog No. 50-290-3695 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A9251100G
In the extracellular space ecto-5AE-nucleotidase (CD73) dephosphorylates adenosine triphosphate (ATP) to produce adenosine. Adenosine has four receptors namely A1R A2AR A2BR and A3R. Adenosine plays a key role in the osteogenic differentiation. A1R induces osteoclast differentiation and A2AR induces osteoblast differentiation.