Learn More
Sigma Aldrich Fine Chemicals Biosciences Adenosine 9925G
Catalog No. 502897725
Encompass
In the extracellular space ecto-5AE-nucleotidase (CD73) dephosphorylates adenosine triphosphate (ATP) to produce adenosine. Adenosine has four receptors namely A1R A2AR A2BR and A3R. Adenosine plays a key role in the osteogenic differentiation. A1R induces osteoclast differentiation and A2AR induces osteoblast differentiation.
Catalog No. 50-289-7725 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A925125G
In the extracellular space ecto-5AE-nucleotidase (CD73) dephosphorylates adenosine triphosphate (ATP) to produce adenosine. Adenosine has four receptors namely A1R A2AR A2BR and A3R. Adenosine plays a key role in the osteogenic differentiation. A1R induces osteoclast differentiation and A2AR induces osteoblast differentiation.