Learn More
Medchemexpress LLC Adenosine A1 receptor activator T62 | 40312-34-3 | 98.9% | 291.80 g/mol | C15H14ClNOS | 1 ML
SDP
Catalog No. 5000242697
Encompass_Preferred
Adenosine A1 receptor activator T62 is a selective positive allosteric modulator of the adenosine A1 receptor supplied for in vitro research. It is available as a solid or as a ready-to-use 10 mM solution in DMSO, with reported purity of 98.93% and a molecular weight of 291.80 g/mol. Follow the SDS for storage and safe handling.
  • Allosteric enhancer of adenosine A1 receptor.
  • Available as solid or 10 mM solution in DMSO.
  • High reported purity suitable for biochemical and cellular assays.
  • High solubility in DMSO (100 mg/mL) for easy formulation.
  • Multilingual SDS provided for safety and handling guidance.
Catalog No. 50-002-42697 Supplier Medchemexpress LLC Supplier No. HY10619910MM 1ML
Adenosine A1 receptor activator T62 is a selective positive allosteric modulator of the adenosine A1 receptor supplied for in vitro research. It is available as a solid or as a ready-to-use 10 mM solution in DMSO, with reported purity of 98.93% and a molecular weight of 291.80 g/mol. Follow the SDS for storage and safe handling.
  • Allosteric enhancer of adenosine A1 receptor.
  • Available as solid or 10 mM solution in DMSO.
  • High reported purity suitable for biochemical and cellular assays.
  • High solubility in DMSO (100 mg/mL) for easy formulation.
  • Multilingual SDS provided for safety and handling guidance.