Learn More
Medchemexpress LLC Adenosine A1 receptor activator T62 | 40312-34-3 | MFCD02738687 | 98.9% | 291.80 | C15H14ClNOS | 1 MG
SDP
Catalog No. 5000244390
Encompass_Preferred
Adenosine A1 receptor activator T62 is an allosteric enhancer of the adenosine A1 receptor used in preclinical research to study A1-mediated signaling and pain modulation. It produces antinociception in animal models of acute, inflammatory, and neuropathic pain.
  • Allosteric enhancer of the adenosine A1 receptor.
  • Demonstrated antinociception in animal models of acute, inflammatory, and neuropathic pain.
  • High purity (~98.9%) suitable for research applications.
  • Solid, light yellow to yellow, suitable for solid or solution preparations.
  • Molecular formula C15H14ClNOS; molecular weight 291.80.
  • Available in small-scale solid amounts and DMSO solution for in vitro use.
Catalog No. 50-002-44390 Supplier Medchemexpress LLC Supplier No. HY1061991MG
Adenosine A1 receptor activator T62 is an allosteric enhancer of the adenosine A1 receptor used in preclinical research to study A1-mediated signaling and pain modulation. It produces antinociception in animal models of acute, inflammatory, and neuropathic pain.
  • Allosteric enhancer of the adenosine A1 receptor.
  • Demonstrated antinociception in animal models of acute, inflammatory, and neuropathic pain.
  • High purity (~98.9%) suitable for research applications.
  • Solid, light yellow to yellow, suitable for solid or solution preparations.
  • Molecular formula C15H14ClNOS; molecular weight 291.80.
  • Available in small-scale solid amounts and DMSO solution for in vitro use.