Learn More
TARGETMOL CHEMICALS INC ADENOSINE AMINE CONGENER 5MG
SDP
Catalog No. 502579160
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Adenosine amine congener (ADAC) (ADAC) is an agonist of selective A1 adenosine receptor. purity: 98%
Catalog No. 50-257-9160 Supplier TARGETMOL CHEMICALS INC Supplier No. T76835MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Adenosine amine congener (ADAC) (ADAC) is an agonist of selective A1 adenosine receptor. purity: 98%