Learn More
Medchemexpress LLC Adenosine amine congener (ADAC) | 96760-69-9 | 99.23% | 25 MG
SDP
Catalog No. 5000338677
Encompass_Preferred
Adenosine amine congener (ADAC) is a selective A1 adenosine receptor agonist. It can ameliorate noise- and cisplatin-induced cochlear injury and demonstrates neuroprotective effects.
  • Selective A1 adenosine receptor agonist
  • Ameliorates noise- and cisplatin-induced cochlear injury
  • Has neuroprotective effects
  • Reduces oxidative stress in noise-exposed cochlea
  • Protects sensory hair cells
  • Reduces cisplatin-induced apoptosis in cochlear tissues
  • Inhibits glutamate release via presynaptic A1 receptors
  • Inhibits voltage-gated Ca2+ channels
  • Mitigates noise-induced hearing loss in a dose- and time-dependent manner
Catalog No. 50-003-38677 Supplier Medchemexpress LLC Supplier No. 25MGHY128064
Adenosine amine congener (ADAC) is a selective A1 adenosine receptor agonist. It can ameliorate noise- and cisplatin-induced cochlear injury and demonstrates neuroprotective effects.
  • Selective A1 adenosine receptor agonist
  • Ameliorates noise- and cisplatin-induced cochlear injury
  • Has neuroprotective effects
  • Reduces oxidative stress in noise-exposed cochlea
  • Protects sensory hair cells
  • Reduces cisplatin-induced apoptosis in cochlear tissues
  • Inhibits glutamate release via presynaptic A1 receptors
  • Inhibits voltage-gated Ca2+ channels
  • Mitigates noise-induced hearing loss in a dose- and time-dependent manner