Learn More
Cell Biolabs Inc Adenosine Assay Kit, 100 assays, Fluorometric, Detection Limit 1.56 μM, 96-well microplate
Catalog No. NC2049490
Encompass_Preferred
The Adenosine Assay Kit provides a simple and reliable fluorometric method to quantify total adenosine levels in various biological samples. It is designed for scientific researchers to investigate the roles of adenosine in energy transfer, cell signaling, and blood flow regulation. The kit is optimized for diverse research applications, allowing detection of adenosine levels as low as 1.56 μM, making it suitable for low-abundance samples.
  • Accurate quantification of total adenosine in biological samples.
  • High sensitivity allows detection of adenosine concentrations as low as 1.56 μM.
  • Compatible with cell lysates, tissue lysates, serum, plasma, and urine samples.
  • Utilizes a 96-well microplate format for efficient, high-throughput analysis.
  • Comprehensive kit includes adenosine standard, assay buffer, fluorometric probe, and necessary enzymes.
Catalog No. NC2049490 Supplier Cell Biolabs Inc Supplier No. MET5090
The Adenosine Assay Kit provides a simple and reliable fluorometric method to quantify total adenosine levels in various biological samples. It is designed for scientific researchers to investigate the roles of adenosine in energy transfer, cell signaling, and blood flow regulation. The kit is optimized for diverse research applications, allowing detection of adenosine levels as low as 1.56 μM, making it suitable for low-abundance samples.
  • Accurate quantification of total adenosine in biological samples.
  • High sensitivity allows detection of adenosine concentrations as low as 1.56 μM.
  • Compatible with cell lysates, tissue lysates, serum, plasma, and urine samples.
  • Utilizes a 96-well microplate format for efficient, high-throughput analysis.
  • Comprehensive kit includes adenosine standard, assay buffer, fluorometric probe, and necessary enzymes.