Learn More
Medchemexpress LLC Adenosine, cyclic 3',5'-(hydrogen phosphate) | 60-92-4 | 99.9% | 1G
SDP
Catalog No. 5000411687
Encompass_Preferred
Cyclic AMP (Cyclic adenosine monophosphate), an adenosine triphosphate derivative, is an intracellular signaling molecule responsible for directing cellular responses to extracellular signals. It acts as an important second messenger in many biological processes.
  • Intracellular signaling molecule for cellular responses to extracellular signals.
  • Modulates mediator generation.
  • Suppresses expression of pro-inflammatory cytokines like TNF-α and IL-12.
  • Enhances production of anti-inflammatory cytokine IL-10.
  • Functions as a microbial and human endogenous metabolite.
Catalog No. 50-004-11687 Supplier Medchemexpress LLC Supplier No. HYB15111G
Cyclic AMP (Cyclic adenosine monophosphate), an adenosine triphosphate derivative, is an intracellular signaling molecule responsible for directing cellular responses to extracellular signals. It acts as an important second messenger in many biological processes.
  • Intracellular signaling molecule for cellular responses to extracellular signals.
  • Modulates mediator generation.
  • Suppresses expression of pro-inflammatory cytokines like TNF-α and IL-12.
  • Enhances production of anti-inflammatory cytokine IL-10.
  • Functions as a microbial and human endogenous metabolite.