Learn More
Medchemexpress LLC Adenosine-d-1 | 119540-53-3 | 99.5% | 5 MG
SDP
Catalog No. 5000406134
Encompass_Preferred
Adenosine-d-1 is a deuterium-labeled version of Adenosine. Adenosine is a naturally occurring autacoid that interacts with four G protein-coupled receptors: A1, A2A, A2B, and A3, influencing various aspects of cellular physiology.
  • This compound can be utilized as a tracer.
  • It can serve as an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06134 Supplier Medchemexpress LLC Supplier No. HYB0228S75MG
Adenosine-d-1 is a deuterium-labeled version of Adenosine. Adenosine is a naturally occurring autacoid that interacts with four G protein-coupled receptors: A1, A2A, A2B, and A3, influencing various aspects of cellular physiology.
  • This compound can be utilized as a tracer.
  • It can serve as an internal standard for quantitative analysis using methods such as NMR, GC-MS, or LC-MS.