Learn More
Medchemexpress LLC Adenosine-d1 | 109923-50-4 | 99.7% | 268.25 g/mol | C10H12DN5O4 | 10 MG
SDP
Catalog No. 5000297068
Encompass_Preferred
Deuterium-labeled adenosine used as a tracer or internal standard for quantitative analytical methods such as LC-MS, GC-MS, and NMR. Supplied as a white to off-white solid with high purity and specified storage conditions to maintain stability.
  • Deuterium-labeled adenosine for tracer and internal standard use.
  • High purity (99.7%) suitable for analytical applications.
  • Soluble in DMSO with ultrasonic warming; compatible with LC-MS, GC-MS, and NMR.
  • Powder stability: up to 3 years at -20°C; follow solvent storage guidelines for solutions.
Catalog No. 50-002-97068 Supplier Medchemexpress LLC Supplier No. HYB0228S10MG
Deuterium-labeled adenosine used as a tracer or internal standard for quantitative analytical methods such as LC-MS, GC-MS, and NMR. Supplied as a white to off-white solid with high purity and specified storage conditions to maintain stability.
  • Deuterium-labeled adenosine for tracer and internal standard use.
  • High purity (99.7%) suitable for analytical applications.
  • Soluble in DMSO with ultrasonic warming; compatible with LC-MS, GC-MS, and NMR.
  • Powder stability: up to 3 years at -20°C; follow solvent storage guidelines for solutions.