Learn More
Medchemexpress LLC Adenosine-d2 | 82741-17-1 | 269.25 | C10H11D2N5O4 | 10 MG
SDP
Catalog No. 5000298990
Encompass_Preferred
Adenosine-d2 is a deuterium-labeled adenosine used as a stable isotope standard and research reagent for analytical and biochemical studies of adenosine-related pathways. It is supplied as a solid (10 mg) and as a 10 mM solution in DMSO, and is commonly used as an internal standard, tracer, or reference compound in mass spectrometry and metabolic studies.
  • Deuterium-labeled analog useful for isotope dilution and tracer experiments.
  • Offered as a 10 mg solid and as a 10 mM solution in DMSO.
  • Molecular weight 269.25 and CAS number 82741-17-1 for unambiguous identification.
  • Suitable for use as an internal standard in LC-MS/MS and metabolic tracing workflows.
Catalog No. 50-002-98990 Supplier Medchemexpress LLC Supplier No. HYB0228S610MG
Adenosine-d2 is a deuterium-labeled adenosine used as a stable isotope standard and research reagent for analytical and biochemical studies of adenosine-related pathways. It is supplied as a solid (10 mg) and as a 10 mM solution in DMSO, and is commonly used as an internal standard, tracer, or reference compound in mass spectrometry and metabolic studies.
  • Deuterium-labeled analog useful for isotope dilution and tracer experiments.
  • Offered as a 10 mg solid and as a 10 mM solution in DMSO.
  • Molecular weight 269.25 and CAS number 82741-17-1 for unambiguous identification.
  • Suitable for use as an internal standard in LC-MS/MS and metabolic tracing workflows.