Learn More
Medchemexpress LLC Adenosine-d2 | 82741-17-1 | 99.6% | 1 ML
SDP
Catalog No. 5000405515
Encompass_Preferred
Adenosine-d2 is the deuterium labeled Adenosine. Adenosine, a ubiquitous endogenous autacoid, acts through the enrollment of four G protein-coupled receptors (A1, A2A, A2B, A3) and affects almost all aspects of cellular physiology.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Isotope-labeled compound
  • Deuterium stable isotope-labeled inhibitor for in vivo cell experiments
  • Relevant for therapeutic drug monitoring (TDM)
Catalog No. 50-004-05515 Supplier Medchemexpress LLC Supplier No. HYB0228S610MM/1ML
Adenosine-d2 is the deuterium labeled Adenosine. Adenosine, a ubiquitous endogenous autacoid, acts through the enrollment of four G protein-coupled receptors (A1, A2A, A2B, A3) and affects almost all aspects of cellular physiology.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Isotope-labeled compound
  • Deuterium stable isotope-labeled inhibitor for in vivo cell experiments
  • Relevant for therapeutic drug monitoring (TDM)