Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Adenosine deaminase 250U | 9026-93-1 | 250 U
SDP

Catalog No. 5000229870
Encompass_Preferred

Adenosine deaminase from calf spleen is a tissue-derived enzyme preparation that catalyzes the irreversible deamination of adenosine and 2'-deoxyadenosine to inosine and 2'-deoxyinosine. Supplied as a defined total activity, it is intended for use in biochemical assays, enzymology, and purine metabolism studies. Manufacturer-provided activity and handling information should be consulted before use.

  • Derived from calf spleen tissue.
  • Provided as 250 U total enzyme activity.
  • Enzyme activity ≥15 U/mg (manufacturer specification).
  • Unit definition: 1 μmol adenosine to inosine per minute at 25°C, pH 7.4.
  • Suitable for biochemical assays and purine metabolism research.
  • Store under conditions specified on the certificate of analysis.

Catalog No. 50-002-29870 Supplier Medchemexpress LLC Supplier No. HY114175A250U
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Adenosine deaminase from calf spleen is a tissue-derived enzyme preparation that catalyzes the irreversible deamination of adenosine and 2'-deoxyadenosine to inosine and 2'-deoxyinosine. Supplied as a defined total activity, it is intended for use in biochemical assays, enzymology, and purine metabolism studies. Manufacturer-provided activity and handling information should be consulted before use.

  • Derived from calf spleen tissue.
  • Provided as 250 U total enzyme activity.
  • Enzyme activity ≥15 U/mg (manufacturer specification).
  • Unit definition: 1 μmol adenosine to inosine per minute at 25°C, pH 7.4.
  • Suitable for biochemical assays and purine metabolism research.
  • Store under conditions specified on the certificate of analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.