Learn More
Abcam Adenosine Deaminase (ADA) Activity Assay Kit, Fluorometric, 100 tests, Detection Method: Fluorescent, Assay Type: Enzyme activity (quantitative), Sensitivity: 10 μU, Assay Range: 10 - 100 μU, Excitation/Emission: 535/587 nm
Catalog No. NC2333751
Encompass_Preferred
Adenosine Deaminase (ADA) Activity Assay Kit, Fluorometric, 100 tests, Detection Method: Fluorescent, Assay Type: Enzyme activity (quantitative), Sensitivity: 10 μU, Assay Range: 10 - 100 μU, Excitation/Emission: 535/587 nm
Catalog No. NC2333751 Supplier Abcam Supplier No. AB204695100TEST
Add to Cart
Add to Cart
Adenosine Deaminase (ADA) Activity Assay Kit, Fluorometric, 100 tests, Detection Method: Fluorescent, Assay Type: Enzyme activity (quantitative), Sensitivity: 10 μU, Assay Range: 10 - 100 μU, Excitation/Emission: 535/587 nm