Learn More
Abcam Adenosine Deaminase (ADA) Activity Assay Kit (Fluorometric)
Supplier:  Abcam AB204695
Encompass_Preferred
Adenosine Deaminase (ADA) Activity Assay Kit (Fluorometric) (ab204695) is an assay where inosine formed from the breakdown of adenosine is detected via a multi-step reaction, resulting in the formation of an intermediate that reacts with the ADA Probe to generate a fluorescent product that can be easily quantified at Ex/Em = 535/587 nm.
Catalog No. NC3877750