Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diazyme Laboratories Adenosine Deaminase Assay

Catalog No. 23700580
Encompass
Encompass_Preferred
Change view
Click to view available options
Type:
Diagnostic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
23-700-580 Diagnostic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-700-580 Supplier Diazyme Laboratories Supplier No. DZ117CK
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Diazyme Adenosine Deaminase (ADA) Assay Kit is for determination of ADA activity in serum and plasma samples.

  • The assay is linear from 0-200 U/L and has precisions of Intra assay %CV < 4.5% and Inter assay %CV < 5.0%
  • The assay is ready-to-use for both manual method and automated chemistry analyzers (Kinetic)
  • The assay is specific for ADA and has no detectable reaction with other nucleosides and is not affected by serum bilirubin up to 30 mg/dL, hemoglobin up to 200 mg/dL, triglycerides up to 750 mg/dL, or ascorbic acid up to 4 mg/dL
  • The assay is based on H2O2detection, which is more sensitive and reliable than the method detecting ammonia production

Specifications

Certifications/Compliance USA: For Research Use only. EU: CE IVD
Content And Storage REAGENT are stable until their expiration date when stored at 2 to 8°C
CE Marker Yes
Concentration Lab dependent
Type Diagnostic
Packaging Type Universal
Form Liquid, Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.