Learn More
Sigma Aldrich Fine Chemicals Biosciences Adenosine Deaminase from b
Supplier:  Sigma Aldrich Fine Chemicals Biosciences A50431KU
Encompass
Molecular weight 32.5-33 kDapI 4.85
Catalog No. 50-290-7162