Learn More
Sigma Aldrich Fine Chemicals Biosciences Adenosine Deaminase from b
Catalog No. 502908363
Encompass
Molecular weight 32.5-33 kDapI 4.85
Catalog No. 50-290-8363 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A51681KU
Molecular weight 32.5-33 kDapI 4.85