Learn More
Bioss Adenosine deaminase Recombinan
SDP
Catalog No. 5000065948
Encompass_Preferred
Adenosine deaminase Primary Antibody Unconjugated Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB IF(ICC) IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol
Catalog No. 50-000-65948 Supplier Bioss Supplier No. BSM62215R
Adenosine deaminase Primary Antibody Unconjugated Host Species Rabbit Target Species Human Rat Clonality Monoclonal Applications WB IF(ICC) IP RecombinantConcentration 1ug/ul Size 100 ul Storage Buffer 10mM phosphate buffered saline(pH 7 4) with 150mM sodium chloride 0 05% BSA 0 02% sodium azide and 50% glycerol